missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-3527091
Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -110.0°C
CAS Min % 99
Color Colorless
CAS Max % 100
Density 0.7404g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 100 mL
Flash Point −28°C
Infrared Spectrum Authentic
Physical Form Liquid
Refractive Index 1.3675 to 1.3695
Assay Percent Range 99% min. (GC)
Residue after Evaporation 0.0005 % max.
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Water 0.005% Max.
Merck Index 156106
Solubility Information Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Formula Weight 88.15
Percent Purity 99+%
Grade Extra Dry
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'