missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl isocyanide, 98%

Product Code. p-7047783
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11399675

Brand: Thermo Scientific Alfa Aesar L19747.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7188-38-7 | C5H9N | 83.134 g/mol

tert-Butyl isocyanide is used in a tin-free procedure for alkyl radical reactions in the presence of a free-radical initiator. It is a useful intermediate in multicomponent reactions. It is also used in the synthesis of coumarines, 4H-chromenes, isoxazolines and to trap 2-cyclopropylidene-1,3-diones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl isocyanide is used in a tin-free procedure for alkyl radical reactions in the presence of a free-radical initiator. It is a useful intermediate in multicomponent reactions. It is also used in the synthesis of coumarines, 4H-chromenes, isoxazolines and to trap 2-cyclopropylidene-1,3-diones.

Solubility
Soluble in organic solvents such as ethanol, methanol, ether, toluene and dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7188-38-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00000002
InChI Key FAGLEPBREOXSAC-UHFFFAOYSA-N
Synonym tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
PubChem CID 23577
IUPAC Name 2-isocyano-2-methylpropane
SMILES CC(C)(C)[N+]#[C-]

Specifications

Density 0.735
Boiling Point 90°C to 92°C
Flash Point −2°C (28°F)
Odor Stench
Refractive Index 1.3755
Quantity 1 g
UN Number UN1992
Beilstein 1903156
Solubility Information Soluble in organic solvents such as ethanol,methanol,ether,toluene and dichloromethane.
Formula Weight 83.13
Percent Purity 98%
Chemical Name or Material tert-Butyl isocyanide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl isocyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : tert-Butyl isocyanide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.