missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl isocyanate, 97%

Product Code. 10461094 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10461094 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10461094 Supplier Thermo Scientific Acros Supplier No. 162960050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1609-86-5 | C5H9NO | 99.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1609-86-5
Density 0.8440g/mL
Boiling Point 84.0°C
Flash Point −4°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C5H9NO
Linear Formula (CH3)3CNCO
Quantity 5 g
Synonym tert-butyl isocyanate, tert-butylisocyanate, propane, 2-isocyanato-2-methyl, t-butyl isocyanate, t-butylisocyanate, unii-2rhq1bs36d, 2-isocyanato-2-methyl-propane, isocyanic acid, tert-butyl ester, 2rhq1bs36d, 1,1-dimethylethyl isocyanate
Solubility Information Solubility in water: reacts.
InChI Key MGOLNIXAPIAKFM-UHFFFAOYSA-N
SMILES CC(C)(C)N=C=O
IUPAC Name 2-isocyanato-2-methylpropane
Molecular Weight (g/mol) 99.133
PubChem CID 62412
Formula Weight 99.13
Percent Purity ≥96.0% (GC)
Chemical Name or Material tert-Butyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.