missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl isocyanate, 97%

Product Code. 10451474 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10451474 100 g Glass bottle 100g
10173434 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10451474 Supplier Thermo Scientific Acros Supplier No. 162961000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1609-86-5 | C5H9NO | 99.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609-86-5
MDL Number MFCD00002035
InChI Key MGOLNIXAPIAKFM-UHFFFAOYSA-N
Synonym tert-butyl isocyanate, tert-butylisocyanate, propane, 2-isocyanato-2-methyl, t-butyl isocyanate, t-butylisocyanate, unii-2rhq1bs36d, 2-isocyanato-2-methyl-propane, isocyanic acid, tert-butyl ester, 2rhq1bs36d, 1,1-dimethylethyl isocyanate
PubChem CID 62412
IUPAC Name 2-isocyanato-2-methylpropane
SMILES CC(C)(C)N=C=O

Specifications

Color Colorless
Density 0.8440g/mL
Boiling Point 84.0°C
Flash Point −4°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C5H9NO
Linear Formula (CH3)3CNCO
Refractive Index 1.3850 to 1.3870 (20°C, 589nm)
Quantity 100 g
Beilstein 04,175
Specific Gravity 0.844
Molecular Weight (g/mol) 99.13
Formula Weight 99.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.