missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydrogen phthalate, 97%

Product Code. 11358215
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358215 5 g 5g
11368215 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358215 Supplier Thermo Scientific Alfa Aesar Supplier No. L08534.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33693-84-4 | C12H14O4 | 222.24 g/mol

It finds its application in the resolution and reactions of tertiary alcohols : 2-phenylbutan-2-ol and 3-methylhexan-3-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the resolution and reactions of tertiary alcohols : 2-phenylbutan-2-ol and 3-methylhexan-3-ol.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33693-84-4
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00043711
InChI Key PBUQZKXKYSAJDO-UHFFFAOYSA-N
Synonym tert-butyl hydrogen phthalate, 2-tert-butoxycarbonyl benzoic acid, unii-8390xkx0sq, 2-tert-butyl oxycarbonyl benzoic acid, t-monobutyl phthalate, t-butyl hydrogen phthalate, acmc-1ad5b, dsstox_cid_27150, dsstox_rid_82154, dsstox_gsid_47150
PubChem CID 134976
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonyl]benzoic acid
SMILES CC(C)(C)OC(=O)C1=CC=CC=C1C(=O)O

Specifications

Melting Point 84°C to 88°C
Quantity 5 g
Beilstein 2263386
Solubility Information Insoluble in water.
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material tert-Butyl hydrogen phthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.