missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl ethyl malonate, 95%

Product Code. 11403914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403914 5 g 5g
11413914 25 g 25g
11423914 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403914 Supplier Thermo Scientific Alfa Aesar Supplier No. A11927.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 32864-38-3 | C9H16O4 | 188.223 g/mol

tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.

Solubility
Miscible with most common solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32864-38-3
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00009193
InChI Key OCOBFMZGRJOSOU-UHFFFAOYSA-N
Synonym tert-butyl ethyl malonate, t-butyl ethyl malonate, propanedioic acid, 1,1-dimethylethyl ethyl ester, 1-tert-butyl 3-ethyl propanedioate, tert-butyl ethyl propanedioate, malonic acid tert-butyl ethyl ester, tert-butyl ethyl propane-1,3-dioate, zlchem 692, acmc-1cjju, 1-tert-butyl ethyl malonate
PubChem CID 96345
IUPAC Name 3-O-tert-butyl 1-O-ethyl propanedioate
SMILES CCOC(=O)CC(=O)OC(C)(C)C

Specifications

Density 0.994
Boiling Point 98°C to 100°C (22 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.416
Quantity 5 g
Beilstein 1776681
Solubility Information Miscible with most common solvents. Immiscible with water.
Formula Weight 188.22
Percent Purity 95%
Chemical Name or Material tert-Butyl ethyl malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.