missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl diethylphosphonoacetate, 95%

Product Code. p-7047116
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388586 1 g 1g
11398586 5 g 5g
11308596 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388586 Supplier Thermo Scientific Alfa Aesar Supplier No. L17651.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 27784-76-5 | C10H21O5P | 252.247 g/mol

tert-Butyl diethylphosphonoacetate is used as a reactant for preparation of hydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents, synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3) and bicyclic triaminophosphine-promoted stereoselective synthesis of a,β-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl diethylphosphonoacetate is used as a reactant for preparation of hydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents, synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3) and bicyclic triaminophosphine-promoted stereoselective synthesis of a,ß-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27784-76-5
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.247
MDL Number MFCD00075414
InChI Key NFEGNISFSSLEGU-UHFFFAOYSA-N
Synonym tert-butyl diethylphosphonoacetate, tert-butyl 2-diethoxyphosphoryl acetate, diethyl boc-methyl phosphonate, diethoxy-phosphoryl-acetic acid tert-butyl ester, t-butyl diethylphosphonoacetate, diethyl phosphonoacetic acid tert-butyl ester, t-butyl diethyl phosphonoacetate, diethylphosphonoacetic acid tert-butyl ester, diethyl 2-tert-butoxycarbonylmethyl phosphonate, acetic acid, diethoxyphosphinyl-, 1,1-dimethylethyl ester
PubChem CID 2773685
IUPAC Name tert-butyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC(C)(C)C)OCC

Specifications

Density 1.074
Boiling Point 100°C to 103°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.431
Quantity 1 g
Beilstein 2050126
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 252.25
Percent Purity 95%
Chemical Name or Material tert-Butyl diethylphosphonoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.