missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Dicarbonate, TRC

Product Code. 30003657
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003657 250 g 250g
30008387 5 g 5g
30008972 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003657 Supplier Toronto Research Chemicals Supplier No. TRCB691045250G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.25
Synonym BOC2O, C,C'-bis(1,1-Dimethylethyl) Ester Dicarbonic Acid, Bis(1,1-dimethylethyl) Ester Dicarbonic Acid, Oxydi-formic Acid Di-tert-butyl Ester, BOC-anhydride, Bis(1,1-dimethylethyl) Dicarbonate, Bis(tert-butyl) Dicarbonate, Di-tert-butyl Dicarbonate, Di-tert-butyl Oxydiformate, Di-tert-butyl Pyrocarbonate, Pyrocarbonic Acid Di-tert-butyl Ester, tert-Butoxycarbonyl Anhydride, tert-Butyl Dicarbonate
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Quantity 250 g
InChI Formula InChI=1S/C10H18O5/c1-9(2,3)14-7(11)13-8(12)15-10(4,5)6/h1-6H3
Formula Weight 218.1154
Chemical Name or Material tert-Butyl Dicarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.