missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl cyanoacetate, 98%

Product Code. 11372665
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372665 100 g 100g
11352665 5 g 5g
11362665 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11372665 Supplier Thermo Scientific Alfa Aesar Supplier No. L00123.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1116-98-9 | C7H11NO2 | 141.17 g/mol

tert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1116-98-9
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00001938
InChI Key BFNYNEMRWHFIMR-UHFFFAOYSA-N
Synonym tert-butyl cyanoacetate, t-butyl cyanoacetate, acetic acid, cyano-, 1,1-dimethylethyl ester, cyanoacetic acid tert-butyl ester, acetic acid, cyano-, tert-butyl ester, acetic acid, 2-cyano-, 1,1-dimethylethyl ester, tert-butylcyanacetat, t-butylcyanoacetate, tert-butylcyanoacetate, tert butyl cyanoacetate
PubChem CID 70693
IUPAC Name tert-butyl 2-cyanoacetate
SMILES CC(C)(C)OC(=O)CC#N

Specifications

Density 0.987
Boiling Point 67°C to 68°C (1.5 mmHg)
Flash Point 91°C (195°F)
Odor Odorless
Refractive Index 1.42
Quantity 100 g
Beilstein 1755933
Solubility Information Difficult to mix.
Formula Weight 141.17
Percent Purity 98%
Chemical Name or Material tert-Butyl cyanoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.