missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl chloroacetate, 98%

Product Code. p-7022871
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432518 25 g 25g
11442518 100 g 100g
11452518 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432518 Supplier Thermo Scientific Alfa Aesar Supplier No. A15833.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-59-5 | C6H11ClO2 | 150.602 g/mol

tert-Butyl chloroacetate is used in the synthesis of imidazol-1-yl-acetic acid hydrochloride, cis-disubstituted aziridine ester via aza-Darzens reaction and 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloroacetate is used in the synthesis of imidazol-1-yl-acetic acid hydrochloride, cis-disubstituted aziridine ester via aza-Darzens reaction and 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore.

Solubility
Decomposes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-59-5
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00000930
InChI Key KUYMVWXKHQSIAS-UHFFFAOYSA-N
Synonym tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate
PubChem CID 66052
IUPAC Name tert-butyl 2-chloroacetate
SMILES CC(C)(C)OC(=O)CCl

Specifications

Density 1.053
Boiling Point 155°C
Flash Point 46°C (115°F)
Odor Ester-like
Linear Formula ClCH2CO2C(CH3)3
Refractive Index 1.423
Quantity 25 g
UN Number UN2929
Beilstein 1753006
Merck Index 14,1563
Solubility Information Decomposes in water.
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material tert-Butyl chloroacetate
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.