missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl chloroacetate, 97%

Product Code. 10254751 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10254751

Brand: Thermo Scientific Acros 347231000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-59-5 | C6H11ClO2 | 150 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-59-5
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150
MDL Number MFCD00000930
InChI Key KUYMVWXKHQSIAS-UHFFFAOYSA-N
Synonym tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate
PubChem CID 66052
IUPAC Name tert-butyl 2-chloroacetate
SMILES CC(C)(C)OC(=O)CCl

Specifications

Color Colorless to Yellow
Density 1.0530g/mL
Boiling Point 48.0°C to 49.0°C (11.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CO2C(CH3)3
Refractive Index 1.4220 to 1.4240
Quantity 100 g
Beilstein 02, III, 444
Fieser 01,86; 03,36
Merck Index 15, 1564
Specific Gravity 1.053
Solubility Information Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in isopropanol,1,2-dichloroethane,toluene
Formula Weight 150
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl chloroacetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl chloroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.