missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Carbazate, TRC

Product Code. 30348378
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348378 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348378 Supplier Toronto Research Chemicals Supplier No. TRCB69217010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl Carbazate
CAS 870-46-2
Molecular Formula C5H12N2O2
Quantity 10 g
Synonym tert-Butyl Ester Carbazic Acid, (tert-Butoxycarbonyl)hydrazide, (tert-Butoxycarbonyl)hydrazine, (tert-Butyloxycarbonyl)hydrazide, 1,1-Dimethylethyl Carbazate, 1,1-Dimethylethyl Hydrazinecarboxylate, 1-(tert-Butoxycarbonyl)hydrazine, Boc-hydrazine, Hydrazinecarboxylic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)hydrazine, N-Boc-Hydrazine, NSC 60250, tert-Butyl N-aminocarbamate, tert-Butyl Carbazate, tert-Butyl Hydrazinecarboxylate, tert-Butyl Hydrazinoformate
InChI Formula InChI=1S/C5H12N2O2/c1-5(2,3)9-4(8)7-6/h6H2,1-3H3,(H,7,8)
SMILES CC(C)(C)OC(=O)NN
IUPAC Name tert-butyl N-aminocarbamate
Molecular Weight (g/mol) 132.16
Formula Weight 132.0899
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.