missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl carbazate, 98+%

Product Code. 11439533
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439533 100 g 100g
11429533 25 g 25g
11449533 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439533 Supplier Thermo Scientific Alfa Aesar Supplier No. A12383.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 870-46-2 | C5H12N2O2 | 132.163 g/mol

tert-Butyl carbazate is used in a palladium-catalyzed cross-coupling with vinyl halides to prepare N-Boc-N-alkenylhydrazines. It is utilized in solid phase peptide synthesis and in the optical purity determination of alfa-amino aldehyde. It reacts with aldehydes to get hydrazones, which finds application as an intermediate in the synthesis of HIV-1 protease inhibitors. In addition to this, it is used as a starting material for the preparation of BOC-azide, sulfonic and carboxylic hydrazides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl carbazate is used in a palladium-catalyzed cross-coupling with vinyl halides to prepare N-Boc-N-alkenylhydrazines. It is utilized in solid phase peptide synthesis and in the optical purity determination of alfa-amino aldehyde. It reacts with aldehydes to get hydrazones, which finds application as an intermediate in the synthesis of HIV-1 protease inhibitors. In addition to this, it is used as a starting material for the preparation of BOC-azide, sulfonic and carboxylic hydrazides.

Solubility
Soluble in ethanol and methanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-46-2
Molecular Formula C5H12N2O2
Molecular Weight (g/mol) 132.163
MDL Number MFCD00007593
InChI Key DKACXUFSLUYRFU-UHFFFAOYSA-N
Synonym tert-butyl carbazate, tert-butyl hydrazinecarboxylate, t-butyl carbazate, hydrazinecarboxylic acid, 1,1-dimethylethyl ester, t-butylcarbazate, tert-butoxycarbonyl hydrazide, boc-hydrazide, boc-hydrazine, tert-butoxycarbonyl hydrazine, carbazic acid tert-butyl ester
PubChem CID 70091
IUPAC Name tert-butyl N-aminocarbamate
SMILES CC(C)(C)OC(=O)NN

Specifications

Melting Point 37°C to 41°C
Boiling Point 63°C to 65°C (0.1 mmHg)
Flash Point 91°C (195°F)
Quantity 100 g
UN Number UN1325
Beilstein 1756967
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol and methanol.
Formula Weight 132.16
Percent Purity ≥98%
Chemical Name or Material tert-Butyl carbazate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.