missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl bromide, 98+%, stab. with potassium carbonate

Product Code. 11454744
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454744 500 g 500g
11444744 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454744 Supplier Thermo Scientific Alfa Aesar Supplier No. A13727.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 507-19-7 | C4H9Br | 137.02 g/mol

tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromide is used as a raw material in synthetic organic chemistry. It is used to study the massive deadenylation of adenine based-nucleosides.

Solubility
Immiscible with water.

Notes
Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-19-7
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000125
InChI Key RKSOPLXZQNSWAS-UHFFFAOYSA-N
Synonym tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane
PubChem CID 10485
IUPAC Name 2-bromo-2-methylpropane
SMILES CC(C)(C)Br

Specifications

Melting Point -20°C
Density 1.21
Boiling Point 74°C to 76°C
Flash Point 18°C (64°F)
Assay Percent Range 98%, stabilized with potassium carbonate
Refractive Index 1.428
Quantity 500 g
UN Number UN2342
Beilstein 1730892
Merck Index 14,1555
Solubility Information Immiscible with water.
Formula Weight 137.02
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material tert-Butyl bromide, stabilized with potassium carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.