missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl alcohol, ACS, 99+%

Product Code. 11347588
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
250mL
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347588 1 L 1 L
11367588 250 mL 250mL
11377588 4 x 1 L 4 L
11357588 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11347588 Supplier Thermo Scientific Alfa Aesar Supplier No. 033278.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-65-0 | C4H10O | 74.123 g/mol

tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient in paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25°C
Density 0.786 g/mL
Boiling Point 83°C
Flash Point 11°C (52°F)
Assay Percent Range >99+%
Refractive Index 1.387
Quantity 1 L
Beilstein 906698
Solubility Information Miscible with water,ethanol,ethyl ether and chloroform.
Formula Weight 74.12
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Tert-Butyl alcohol, For ACS analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl alcohol, ACS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: tert-Butyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.