missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Acrylate (Stabilized with MEHQ), TRC

Product Code. 30330255
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330255 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330255 Supplier Toronto Research Chemicals Supplier No. TRCB690506100ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl Acrylate
CAS 1663-39-4
Molecular Formula C7H12O2
Quantity 100 mL
Synonym 1,1-Dimethylethyl 2-Propenoate, 2-Propenoic Acid tert-Butyl Ester, NSC 20950, tert-Butyl 2-Propenoate, tert-Butyl Acrylate, tert-Butyl Propenoate
InChI Formula InChI=1S/C7H12O2/c1-5-6(8)9-7(2,3)4/h5H,1H2,2-4H3
SMILES CC(C)(C)OC(=O)C=C
IUPAC Name tert-butyl prop-2-enoate
Molecular Weight (g/mol) 128.17
Formula Weight 128.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.