missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl acrylate, 99%, stabilized

Product Code. 10247543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
100mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10247543 5 L Plastic drum 5 L
10631663 100 mL Glass bottle 100mL
10161052 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10247543 Supplier Thermo Scientific Acros Supplier No. 371130051
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1663-39-4 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1663-39-4 , 150-76-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00008809
InChI Key ISXSCDLOGDJUNJ-UHFFFAOYSA-N
Synonym tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
PubChem CID 15458
IUPAC Name tert-butyl prop-2-enoate
SMILES CC(C)(C)OC(=O)C=C

Specifications

Melting Point -69.0°C
Density 0.8830g/mL
Boiling Point 121.0°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula H2C=CHCO2C(CH3)3
Refractive Index 1.4100 to 1.4120
Quantity 5 L
Beilstein 02, IV, 1465
Specific Gravity 0.883
Solubility Information Solubility in water: 2g/L
Stabilizer 10 to 20ppm MEHQ
Viscosity 0.9 mPa.s (20°C)
Formula Weight 128.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material tert-Butyl acrylate, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl acrylate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.