missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl acetoacetate, 97%

Product Code. p-79244 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10366120 100 mL 100mL
10020540 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10366120 Supplier Thermo Scientific Supplier No. 107781000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1694-31-1 | C8H14O3 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1694-31-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.2
MDL Number MFCD00008811
InChI Key JKUYRAMKJLMYLO-UHFFFAOYSA-N
Synonym tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
PubChem CID 15538
IUPAC Name tert-butyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OC(C)(C)C

Specifications

Melting Point -38°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 190°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2CO2C(CH3)3
Refractive Index 1.4190 to 1.421
Quantity 100 mL
Fieser 01,83
Specific Gravity 0.97
Solubility Information Solubility in water: 9g/L water (20°C). Other solubilities: soluble in xylene,miscible with most organic solvents
Formula Weight 158.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material tert-Butyl acetoacetate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.