missing translation for 'onlineSavingsMsg'
Learn More

Tert-butyl Acetate, TRC

Product Code. 30450411
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450411 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450411 Supplier Toronto Research Chemicals Supplier No. TRCT88648010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetic acid-tert.-butyl ester
CAS 540-88-5
Molecular Formula C6 H12 O2
Quantity 10 g
Synonym Acetic acid','tert-butyl ester (8CI), tert-Butyl alcohol','acetate (6CI), 1,1-Dimethylethyl acetate, 2-Acetoxy-2-methylpropane, Acetic acid tert-butyl ester, NSC 59719, TBAc, Texaco lead appreciator, tert-Butyl acetate, tert-Butyl ethanoate
InChI Formula InChI=1S/C6H12O2/c1-5(7)8-6(2,3)4/h1-4H3
SMILES CC(=O)OC(C)(C)C
IUPAC Name tert-butyl acetate
Molecular Weight (g/mol) 116.16
Formula Weight 116.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.