missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl acetate, 99%

Product Code. 11312756
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312756 250 g 250g
11302756 50 g 50g
11322756 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11312756 Supplier Thermo Scientific Alfa Aesar Supplier No. L08855.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-88-5 | C6H12O2 | 116.16 g/mol

tert-Butyl acetate is used as a solvent in the production of lacquers, enamels, inks, adhesives, thinners, industrial cleaners and coating blends It is also used as a process solvent for chemical reactions and polymerization. In synthetic organic chemistry, it is used as a tert-butyl group. It serves as an intermediate in the production of pharmaceuticals and synthetic flavorings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl acetate is used as a solvent in the production of lacquers, enamels, inks, adhesives, thinners, industrial cleaners and coating blends It is also used as a process solvent for chemical reactions and polymerization. In synthetic organic chemistry, it is used as a tert-butyl group. It serves as an intermediate in the production of pharmaceuticals and synthetic flavorings.

Solubility
Miscible with ether and ethanol. Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-88-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008807
InChI Key WMOVHXAZOJBABW-UHFFFAOYSA-N
Synonym t-butyl acetate, acetic acid tert-butyl ester, tert-butyl ethanoate, acetic acid, 1,1-dimethylethyl ester, texaco lead appreciator, acetic acid, tert-butyl ester, tert-butylacetate, tert butyl acetate, 1,1-dimethylethyl acetate, 1,1-dimethyl acetate
PubChem CID 10908
IUPAC Name tert-butyl acetate
SMILES CC(=O)OC(C)(C)C

Specifications

Melting Point -62°C
Density 0.863
Boiling Point 94°C to 96°C
Flash Point 4°C (39°F)
Linear Formula CH3CO2C(CH3)3
Refractive Index 1.3865
Quantity 250 g
UN Number UN1123
Beilstein 1699506
Merck Index 14,1537
Solubility Information Miscible with ether and ethanol. Immiscible with water.
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material tert-Butyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.