missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 6-fluoro-1H-pyrazolo[3,4-b]pyridine-3-carboxylate, ≥97%, Thermo Scientific™

Product Code. 10045102
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045102 1 g Amber Glass Bottle 1g
10117392 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10045102 Supplier Thermo Scientific Maybridge Supplier No. MO07504DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 155601-71-1
Molecular Formula C11H12FN3O2
Molecular Weight (g/mol) 237.234
MDL Number MFCD06797502
InChI Key INGAFDNQIOIZHC-UHFFFAOYSA-N
Synonym tert-butyl 6-fluoro-1h-pyrazolo 3,4-b pyridine-3-carboxylate, 6-fluoro-1h-pyrazolo 3,4-b pyridine-3-carboxylic acid tert-butyl ester, tert-butyl6-fluoro-1h-pyrazolo 3,4-b pyridine-3-carboxylate, 3-tert-butoxycarbonyl-6-fluoro-1h-pyrazolo 3,4-b pyridine, tert-butyl 6-fluoro-2h-pyrazolo 3,4-b pyridine-3-carboxylate, t-butyl 6-fluoro-1h-pyrazolo 3,4-b pyridine-3-carboxylate, tert-butyl 6-fluoropyrazolo 5,4-b pyridine-3-carboxylate, tert-butyl 6-fluoro-1h-pyrazolo-3,4-b pyridine-3-carboxylate, tert-butyl 6-fluoro-1h-pyrazolo 3.4-b pyridine-3-carboxylate
PubChem CID 7127829
IUPAC Name tert-butyl 6-fluoro-2H-pyrazolo[3,4-b]pyridine-3-carboxylate
SMILES CC(C)(C)OC(=O)C1=C2C=CC(=NC2=NN1)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.23
Percent Purity 97%
Chemical Name or Material tert-Butyl 6-fluoro-1H-pyrazolo[3, 4-b]pyridine-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.