missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10366843
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366843 1 g Amber Glass Bottle 1g
10357584 250 mg Amber Glass Bottle 250mg
11356835 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366843 Supplier Thermo Scientific Maybridge Supplier No. CC53239DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 864771-44-8
Molecular Formula C18H25BN2O4
Molecular Weight (g/mol) 344.22
MDL Number MFCD08437629
InChI Key MMXDKXUKSKMROG-UHFFFAOYSA-N
Synonym tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-boc-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-n-boc-indazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-indazole-1-carboxylic acid tert-butyl ester, 1-boc-indazole-5-boronic acid pinacol ester, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate, 1-tert-butoxycarbonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1h-indazole-1-carboxylic acid, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate
PubChem CID 18525862
IUPAC Name tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1N=CC2=CC(=CC=C12)B1OC(C)(C)C(C)(C)O1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 344.22
Percent Purity 97%
Chemical Name or Material tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.