missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate, 97%, Thermo Scientific™

Catalog Number 10357584
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
864771-44-8
C18H25BN2O4
344.22
MFCD08437629
MMXDKXUKSKMROG-UHFFFAOYSA-N
tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-boc-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-n-boc-indazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-indazole-1-carboxylic acid tert-butyl ester, 1-boc-indazole-5-boronic acid pinacol ester, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate, 1-tert-butoxycarbonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1h-indazole-1-carboxylic acid, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate
18525862
tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate
CC(C)(C)OC(=O)N1N=CC2=CC(=CC=C12)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
864771-44-8
C18H25BN2O4
344.22
MFCD08437629
MMXDKXUKSKMROG-UHFFFAOYSA-N
tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-boc-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-n-boc-indazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-indazole-1-carboxylic acid tert-butyl ester, 1-boc-indazole-5-boronic acid pinacol ester, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate, 1-tert-butoxycarbonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1h-indazole-1-carboxylic acid, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate
18525862
tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate
CC(C)(C)OC(=O)N1N=CC2=CC(=CC=C12)B1OC(C)(C)C(C)(C)O1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 344.22
Percent Purity 97%
Chemical Name or Material tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'