missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10402273
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10402273 1 g 1g
10413214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402273 Supplier Thermo Scientific Maybridge Supplier No. CC31904DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 844891-13-0
Molecular Formula C11H16N2O3
Molecular Weight (g/mol) 224.26
MDL Number MFCD06659076
InChI Key AZJWODKWFMBMCM-UHFFFAOYSA-N
PubChem CID 2795543
IUPAC Name tert-butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate
SMILES CC1=NN(C(=O)OC(C)(C)C)C(C)=C1C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym tert-butyl 4-formyl-3,5-dimethyl-1h-pyrazole-1-carboxylate, tert-butyl4-formyl-3,5-dimethyl-1h-pyrazole-1-carboxylate, tert-butyl 4-formyl-3,5-dimethylpyrazolecarboxylate, 3,5-dimethyl-4-formyl-1h-pyrazole, n1-boc protected, 1h-pyrazole-1-carboxylicacid, 4-formyl-3,5-dimethyl-, 1,1-dimethylethyl ester
Formula Weight 224.26
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-formyl-3, 5-dimethyl-1H-pyrazole-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.