missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%

Product Code. 15450766
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450766 250 mg 250mg
15430766 1 g 1g
15440766 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450766 Supplier Thermo Scientific Alfa Aesar Supplier No. H54100.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 114772-40-6 | C18H19BrO2 | 347.252 g/mol

1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

Solubility
Soluble in chloroform, ethyl acetate, and DMSO.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-40-6
Molecular Formula C18H19BrO2
Molecular Weight (g/mol) 347.252
MDL Number MFCD06657561
InChI Key YHXCWNQNVMAENQ-UHFFFAOYSA-N
Synonym 2-boc-4'-bromomethyl biphenyl, tert-butyl 4'-bromomethyl biphenyl-2-carboxylate, tert-butyl 2-4-bromomethyl phenyl benzoate, tert-butyl 4'-bromomethyl-1,1'-biphenyl-2-carboxylate, unii-me3i28o0tc, tert-butyl 4'-bromomethyl-2-biphenylcarboxylate, me3i28o0tc, telmisartan impurity h, tert-butyl 2-4-bromomethylphenyl benzoate, 1,1-dimethylethyl 4'-bromomethyl biphenyl-2-carboxylate
PubChem CID 9906257
IUPAC Name tert-butyl 2-[4-(bromomethyl)phenyl]benzoate
SMILES CC(C)(C)OC(=O)C1=CC=CC=C1C2=CC=C(C=C2)CBr

Specifications

Quantity 250 mg
Solubility Information Soluble in chloroform,ethyl acetate,and DMSO.
Formula Weight 347.24
Percent Purity 97%
Chemical Name or Material tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.