missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(Aminomethyl)indoline-1-carboxylate, TRC

Product Code. 30340473
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340473 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340473 Supplier Toronto Research Chemicals Supplier No. TRCB81138810MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl 4-(Aminomethyl)indoline-1-carboxylate
CAS 1086392-32-6
Molecular Formula C14H20N2O2
Quantity 10 mg
InChI Formula InChI=1S/C14H20N2O2/c1-14(2,3)18-13(17)16-8-7-11-10(9-15)5-4-6-12(11)16/h4-6H,7-9,15H2,1-3H3
SMILES CC(C)(C)OC(=O)N1CCc2c(CN)cccc12
IUPAC Name tert-butyl 4-(aminomethyl)-2,3-dihydroindole-1-carboxylate
Molecular Weight (g/mol) 248.32
Formula Weight 248.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.