missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(5-iodopyrid-2-yl)piperazine-1-carboxylate, ≥97%, Thermo Scientific™

Product Code. 10341063
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10341063 1 g 1g
10382195 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10341063 Supplier Thermo Scientific Maybridge Supplier No. CC43911DA
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 497915-42-1
Molecular Formula C14H20IN3O2
Molecular Weight (g/mol) 389.24
MDL Number MFCD07781193
InChI Key RJHWOWJRQNENIC-UHFFFAOYSA-N
Synonym 1-boc-4-5-iodo-2-pyridyl piperazine, tert-butyl 4-5-iodopyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-5-iodopyrid-2-yl piperazine-1-carboxylate, 1-boc-4-5-iodopyridin-2-yl piperazine, 4-5-iodopyridin-2-yl piperazine-1-carboxylic acid tert-butyl ester, acmc-1aoz2, tert-butyl 4-5-iodanylpyridin-2-yl piperazine-1-carboxylate, 4-5-iodo-2-pyridinyl-1-piperazinecarboxylic acid tert-butyl ester, 1-piperazinecarboxylicacid,4-5-iodo-2-pyridinyl-,1,1-dimethylethyl ester
PubChem CID 7164639
IUPAC Name tert-butyl 4-(5-iodopyridin-2-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(I)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 389.23
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-(5-iodopyrid-2-yl)piperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.