missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate, ≥97%, Thermo Scientific™

Product Code. p-3174930
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10587273 250 mg Amber Glass Bottle 250mg
10321043 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10587273 Supplier Thermo Scientific Maybridge Supplier No. CC31109CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-66-0
Molecular Formula C15H24N2O3S
Molecular Weight (g/mol) 312.428
MDL Number MFCD07368658
InChI Key YNLQKGMVENCCLE-UHFFFAOYSA-N
Synonym tert-butyl 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridinecarboxylate, tert-butyl 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine-1-carboxylate, 1-piperidinecarboxylicacid, 4-5-hydroxymethyl-4-methyl-2-thiazolyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine, 4-5-hydroxymethyl-4-methyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine, n1-boc protected, tert-butyl 4-5-hydroxymethyl-4-methylthiazol-2-yl piperidine-1-carboxylate
PubChem CID 7164575
IUPAC Name tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate
SMILES CC1=C(SC(=N1)C2CCN(CC2)C(=O)OC(C)(C)C)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 312.43
Percent Purity 95%
Chemical Name or Material Tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.