missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(5-formylpyrid-2-yl)piperazine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10172894
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10172894

Brand: Thermo Scientific Maybridge CC43904DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 479226-10-3
Molecular Formula C15H21N3O3
Molecular Weight (g/mol) 291.351
MDL Number MFCD08271923
InChI Key NETKWTAEPIGRLQ-UHFFFAOYSA-N
PubChem CID 18525767
IUPAC Name tert-butyl 4-(5-formylpyridin-2-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym tert-butyl 4-5-formylpyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-5-formylpyrid-2-yl piperazine-1-carboxylate, 4-5-formylpyridin-2-yl piperazine, 4-5-formylpyridin-2-yl piperazine, n1-boc protected, 1-piperazinecarboxylicacid, 4-5-formyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-formyl-2-pyridinyl-,1,1-dimethylethyl ester
Formula Weight 291.35
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-(5-formylpyrid-2-yl)piperazine-1-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.