missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(4-formylbenzyl)piperazine-1-carboxylate, 95%, Thermo Scientific™

Product Code. 10035762
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10035762 1 g 1g
10360993 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10035762 Supplier Thermo Scientific Maybridge Supplier No. CC30304DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 844891-09-4
Molecular Formula C17H24N2O3
Molecular Weight (g/mol) 304.39
InChI Key UMXKYTSTYWRBBK-UHFFFAOYSA-N
Synonym tert-butyl 4-4-formylbenzyl piperazine-1-carboxylate, tert-butyl 4-4-formylphenyl methyl piperazine-1-carboxylate, 4-piperazin-4-yl methyl benzaldehyde, n1-boc protected, tert-butyl 4-4-formylbenzyl-1-piperazinecarboxylate, tert-butyl-4-4-formylbenzyl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-4-formylphenyl methyl-, 1,1-dimethylethyl ester
PubChem CID 2795517
IUPAC Name tert-butyl 4-[(4-formylphenyl)methyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 304.39
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(4-formylbenzyl)piperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.