missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate, ≥97%, Thermo Scientific™

Product Code. 10619764
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10619764 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10619764 Supplier Thermo Scientific Maybridge Supplier No. CC30016DA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 186650-98-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C16H21N3O2
MDL Number MFCD03791198
Quantity 1 g
Synonym tert-butyl 4-4-cyanophenyl piperazine-1-carboxylate, 1-boc-4-4-cyanophenyl piperazine, tert-butyl 4-4-cyanophenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-piperazinecarboxylic acid, 4-4-cyanophenyl-, 1,1-dimethylethyl ester, 4-piperazin-4-yl benzonitrile,n1-boc protected, n-tert-butoxycarbonyl-4-4-cyanophenyl piperazine, t-butyl 4-4-cyanophenyl piperazine-1-carboxylate, 4-4-cyanophenyl piperazine-1-carboxylic acid tert-butyl ester, 4-4-cyano-phenyl-piperazine-1-carboxylic acid tert-butyl ester
InChI Key WIBWVYVIBJOVIW-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C#N
IUPAC Name tert-butyl 4-(4-cyanophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol) 287.363
PubChem CID 2795512
Formula Weight 287.36
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.