missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥90%, Thermo Scientific™

Product Code. p-3174915
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10027512 1 g 1g
10158414 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10027512 Supplier Thermo Scientific Maybridge Supplier No. CC30013DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Identificadores de productos químicos

CAS 852180-47-3
Molecular Formula C16H25N3O2
Molecular Weight (g/mol) 291.40
MDL Number MFCD03791213
InChI Key TTXMFUXVXBAVIP-UHFFFAOYSA-N
Synonym 1-boc-4-4-aminomethylphenyl piperazine, tert-butyl 4-4-aminomethyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-aminomethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-4-aminomethyl-phenyl-piperazine, 1-4-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 4-4-tert-butoxycarbonyl piperazin-1-yl benzylamine, n1-boc-4-aminomethyl phenylpiperazine, 4-aminomethyl phenylpiperazine, n1-boc protected, 1-4-aminomethyl-phenyl-piperazine-4-carboxylic a
PubChem CID 2763841
IUPAC Name tert-butyl 4-[4-(aminomethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(CN)C=C1

Especificaciones

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 291.39
Percent Purity 90%
Chemical Name or Material Tert-butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.