missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Quantity | 5 g |
Formula Weight | 291.39 |
Percent Purity | 90% |
Chemical Name or Material | Tert-butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'