missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate, 97%, Maybridge™

Product Code. 10182844
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10182844 5 g 5g
10482444 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10182844 Supplier Thermo Scientific Maybridge Supplier No. MO08404DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1048970-17-7
Molecular Formula C16H30BNO4
Molecular Weight (g/mol) 311.23
MDL Number MFCD09997792
InChI Key IBLQMWKHENBVJE-UHFFFAOYSA-N
Synonym tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine-1-carboxylate, n-boc-piperidine-4-boronic acid pinacol ester, 1-boc-piperidin-4-ylboronic acid pinacol ester, 1-boc-piperidine-4-boronic acid pinacol ester, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine-1-carboxylate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-piperidine-1-carboxylate, 1-boc-piperidin-4-ylboronic acid, pinacol ester, tert-butoxycarbonylpiperidine-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl piperidine, pubchem21000
PubChem CID 43811073
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 311.23
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.