missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate, 97%, Thermo Scientific™

Product Code. p-3629747
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10305904 1 g 1g
10341244 5 g 5g
10056944 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10305904 Supplier Thermo Scientific Maybridge Supplier No. MO08253DA
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 286961-14-6
Molecular Formula C16H28BNO4
Molecular Weight (g/mol) 309.21
MDL Number MFCD03840345
InChI Key VVDCRJGWILREQH-UHFFFAOYSA-N
Synonym n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, 3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester, n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine, tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate
PubChem CID 4642098
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=CC1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 309.21
Percent Purity 97%
Chemical Name or Material N-(tert-Butoxycarbonyl)-3, 6-dihydro-2H-pyridine-4-boronic acidpinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.