missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(3-hydroxyphenyl)perhydro-1,4-diazepine-1-carboxylate, 95%, Thermo Scientific™

Product Code. 10172794
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10172794 250 mg Amber Glass Bottle 250mg
10086722 1 g Amber Glass Bottle 1g
11346895 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10172794 Supplier Thermo Scientific Maybridge Supplier No. MO07901CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-68-9
Molecular Formula C16H24N2O3
Molecular Weight (g/mol) 292.38
MDL Number MFCD08690329
InChI Key QDERCJJDCUXJSL-UHFFFAOYSA-N
Synonym tert-butyl 4-3-hydroxyphenyl perhydro-1,4-diazepine-1-carboxylate, tert-butyl 4-3-hydroxyphenyl-1,4-diazepane-1-carboxylate, 1h-1,4-diazepine-1-carboxylicacid, hexahydro-4-3-hydroxyphenyl-, 1,1-dimethylethyl ester, 1h-1,4-diazepine-1-carboxylicacid,hexahydro-4-3-hydroxyphenyl-,1,1-dimethylethyl ester
PubChem CID 18525947
IUPAC Name tert-butyl 4-(3-hydroxyphenyl)-1,4-diazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCN(CC1)C1=CC(O)=CC=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 292.38
Percent Purity 95%
Chemical Name or Material Tert-butyl 4-(3-hydroxyphenyl)perhydro-1, 4-diazepine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.