missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10642875
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10642875 5 g 5g
10780364 250 mg 250mg
10506713 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10642875 Supplier Thermo Scientific Maybridge Supplier No. CC30414DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 361345-40-6
Molecular Formula C16H25N3O2
Molecular Weight (g/mol) 291.395
InChI Key VKSINZNBWVJILC-UHFFFAOYSA-N
Synonym tert-butyl 4-3-aminobenzyl piperazine-1-carboxylate, 1-boc-4-3-aminobenzyl piperazine, 3-4-boc-piperazin-1-yl-methyl aniline, tert-butyl 4-3-aminophenyl methyl piperazine-1-carboxylate, 3-1-boc-piperazin-4-yl-methyl-aniline, tert-butyl 4-3-aminophenyl methyl piperazinecarboxylate, 4-3-amino-benzyl-piperazine-1-carboxylic acid tert-butyl ester, pubchem12209, acmc-1ah36, 4-3-aminobenzyl piperazine, n1-boc protected
PubChem CID 7127799
IUPAC Name tert-butyl 4-[(3-aminophenyl)methyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC2=CC(=CC=C2)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 291.39
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(3-aminobenzyl)piperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.