missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(2-(aminomethyl)phenyl)piperazine-1-carboxylate, ≥97%, Thermo Scientific™

Product Code. 10117722
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10117722 1 g Amber Glass Bottle 1g
10085822 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10117722 Supplier Thermo Scientific Maybridge Supplier No. CC30713DA
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 174855-53-9
Molecular Formula C16H25N3O2
Molecular Weight (g/mol) 291.395
MDL Number MFCD04115066
InChI Key UINNZXQKTNAOBL-UHFFFAOYSA-N
Synonym tert-butyl 4-2-aminomethyl phenyl piperazine-1-carboxylate, 1-boc-4-2-aminomethyl phenyl piperazine, 1-boc-4-2-aminomethyl-phenyl-piperazine, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 1-piperazinecarboxylicacid, 4-2-aminomethyl phenyl-, 1,1-dimethylethyl ester, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic a, 2-4-t-butoxycarbonylpiperazin-1-yl phenylmethylamine, 4-2-aminomethyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-aminomethyl phenyl-1-piperazinecarboxylic acid tert-butyl ester
PubChem CID 4961274
IUPAC Name tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 291.39
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.