missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3S)-3-aminobutanoate, 95%, Thermo Scientific Chemicals

Product Code. 10287263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10287263 1 g 1g
10728552 100 mg 100mg
10595223 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10287263 Supplier Thermo Scientific Acros Supplier No. 337940010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161105-54-0
MDL Number MFCD00798294
InChI Key BFFNZGWJTHWUMY-LURJTMIESA-N
PubChem CID 7021083
IUPAC Name tert-butyl (3S)-3-aminobutanoate
SMILES CC(CC(=O)OC(C)(C)C)N

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Molecular Formula C8H17NO2
Packaging Glass Bottle
Refractive Index 1.4230 to 1.427
Specific Rotation + 24
Synonym tert-butyl 3s-3-aminobutanoate, s-tert-butyl 3-aminobutanoate, butanoic acid, 3-amino-, 1,1-dimethylethyl ester, 3s, tert-butyl-3s-3-aminobutanoate, tert-butyl s-3-aminobutyrate, 1,1-dimethylethyl 35-3-aminobutanoate, s-3-aminobutyric acid tert-butyl ester, 1,1-dimethylethyl 3s-3-aminobutanoate, butanoic acid,3-amino-, 1,1-dimethylethyl ester, 3s
Specific Rotation Condition + 24.00 (20.00°C c=1,CHCl3)
Molecular Weight (g/mol) 159.23
Quantity 1 g
Formula Weight 159.23
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material tert-Butyl (3S)-3-aminobutanoate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (3S)-3-aminobutanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.