missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3S)-3-amino-4-phenylbutanoate, 97%

Product Code. 10746052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10746052 100 mg Glass Bottle 100mg
10265571 500 mg Glass Bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10746052 Supplier Thermo Scientific Acros Supplier No. 344461000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120686-17-1 | C14H22NO2 | 236.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120686-17-1
Molecular Formula C14H22NO2
Molecular Weight (g/mol) 236.33
MDL Number MFCD00798308
InChI Key ZIJHIHDFXCNFAA-GFCCVEGCSA-O
PubChem CID 1501868
IUPAC Name tert-butyl (3S)-3-amino-4-phenylbutanoate
SMILES CC(C)(C)OC(=O)C[C@H]([NH3+])CC1=CC=CC=C1

Specifications

Color White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Specific Rotation 2.3
Synonym s-tert-butyl 3-amino-4-phenylbutanoate, tert-butyl 3s-3-amino-4-phenylbutanoate, t-butyl 3s-3-amino-4-phenylbutanoate, 1,1-dimethylethyl 3s-3-amino-4-phenylbutanoate, benzenebutanoic acid, b-amino-, 1,1-dimethylethyl ester, bs, t-butyl 3r-3-amino-4-phenylbutanoate, s-tert-butyl3-amino-4-phenylbutanoate, tert-butyl 3s-3-amino-4-phenyl-butanoate, s-3-amino-4-phenyl-butyric acid tert-butyl ester, s-3-amino-4-phenylbutanoic acid tert-butyl ester
Quantity 100 mg
Formula Weight 235.3
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material tert-Butyl (3S)-3-amino-4-phenylbutanoate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (3S)-3-amino-4-phenylbutanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.