missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl (3R)-3-amino-5-methylhexanoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
Specifications
Color | Colorless to Yellow |
Assay Percent Range | 96% min. (GC) |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Synonym | tert-butyl 3r-3-amino-5-methylhexanoate, hexanoicacid, 3-amino-5-methyl-, 1,1-dimethylethyl ester, 3r, tert-butyl 3r-3-amino-5-methyl-hexanoate, r-tert-butyl 3-amino-5-methylhexanoate, 3r-3-amino-5-methylhexanoic acid tert-butyl ester, hexanoicacid,3-amino-5-methyl-,1,1-dimethylethyl ester, 3r |
Refractive Index | 1.4300 to 1.432 |
Specific Rotation | -15 |
Formula Weight | 201.31 |
Percent Purity | 97% |
Specific Rotation Condition | - 15.00 (20.00°C c=1,CHCl3) |
Show More |
Safety and Handling
Product Identifier
- tert-Butyl (3R)-3-amino-5-methylhexanoate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'