missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%

Product Code. 10057592 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10057592 250 mg 250mg
10464415 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10057592 Supplier Thermo Scientific Acros Supplier No. 337882500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 161671-34-7 | C13H19NO2 | 221.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161671-34-7
Molecular Weight (g/mol) 221.3
MDL Number MFCD00798309
InChI Key TYYCBAISLMKLMT-LLVKDONJSA-N
Synonym tert-butyl 3r-3-amino-3-phenylpropanoate, r-tert-butyl 3-amino-3-phenylpropanoate, 3r-3-phenyl-beta-alanine tert-butyl ester, 1,1-dimethylethyl 3r-3-amino-3-phenylpropanoate, tert-butyl-3r-3-amino-3-phenylpropanoate, benzenepropanoic acid,b-amino-,1,1-dimethylethyl ester, br, benzenepropanoic acid, b-amino-, 1,1-dimethylethyl ester, br
PubChem CID 2733824
IUPAC Name tert-butyl (3R)-3-amino-3-phenylpropanoate
SMILES CC(C)(C)OC(=O)CC(C1=CC=CC=C1)N

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C13H19NO2
Packaging Glass Bottle
Refractive Index 1.4960 to 1.498
Specific Rotation + 17
Specific Rotation Condition + 17.00 (20.00°C c=1,CHCl3)
Quantity 250 mg
Formula Weight 221.3
Percent Purity 97%
Physical Form Slightly Viscous Liquid
Chemical Name or Material tert-Butyl (3R)-3-amino-3-phenylpropanoate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (3R)-3-amino-3-phenylpropanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.