missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-(thiocarbamoyl)piperidine-1-carboxylate, 95%, Thermo Scientific™

Product Code. 11346905
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11346905 5 g Amber Glass Bottle 5g
10672275 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346905 Supplier Thermo Scientific Maybridge Supplier No. MO08288DE
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 274682-80-3
Molecular Formula C11H20N2O2S
Molecular Weight (g/mol) 244.353
MDL Number MFCD10700060
InChI Key ZCELPXRDWJCIJE-UHFFFAOYSA-N
Synonym tert-butyl 3-thiocarbamoyl piperidine-1-carboxylate, 1-tert-butoxycarbonyl piperidine-3-thiocarboxamide, 3-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester, 1-tert-butoxycarbonyl-3-carbamothioylpiperidine, 3-carbamothioylpiperidine, n1-boc protected, 3-carbamothioylpiperidine,n1-boc protected, tert-butyl3-carbamothioylpiperidine-1-carboxylate, tert-butyl 3-aminothioxomethyl piperidinecarboxylate, tert-butyl 3-aminocarbonothioyl piperidine-1-carboxylate, 1-piperidinecarboxylicacid, 3-aminothioxomethyl-, 1,1-dimethylethyl ester
PubChem CID 23546083
IUPAC Name tert-butyl 3-carbamothioylpiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=S)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 244.36
Percent Purity 95%
Chemical Name or Material Tert-butyl 3-(thiocarbamoyl)piperidine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.