missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, 97%, Thermo Scientific™

Product Code. p-4510199
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10065682

Brand: Thermo Scientific Maybridge MO07066DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 185099-67-6
Molecular Formula C12H19NO3
Molecular Weight (g/mol) 225.288
InChI Key MENILFUADYEXNU-UHFFFAOYSA-N
Synonym tert-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-boc-nortropinone, n-boc-4-nortropinone, n-tert-butoxycarbonyl nortropinone, 8-boc-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-aza-bicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester, t-butyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid tert-butyl ester, n-boc-nororopinone, 8-tert-butoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one;
PubChem CID 2794767
IUPAC Name tert-butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CC(C)(C)OC(=O)N1C2CCC1CC(=O)C2

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 225.29
Percent Purity 97%
Chemical Name or Material tert-Butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.