missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-(hydroxymethyl)-1-indolinecarboxylate, 97%, Thermo Scientific™

Product Code. 10082782
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10082782 1 g 1g
10505113 250 mg 250mg
10041294 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10082782 Supplier Thermo Scientific Maybridge Supplier No. MO01134DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 172647-87-9
Molecular Formula C14H19NO3
Molecular Weight (g/mol) 249.31
MDL Number MFCD06411549
InChI Key JNQNQSCDMKNYEX-UHFFFAOYSA-N
Synonym tert-butyl 2-hydroxymethyl-1-indolinecarboxylate, 1-t-butoxycarbonyl-2-hydroxymethylindoline, tert-butyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1-boc-2,3-dihydro-2-hydroxymethyl-1h-indole, tert-butyl 2-hydroxymethyl indoline-1-carboxylate, tert-butyl 2-hydroxymethyl indolinecarboxylate, tert-butyl 2-hydroxymethyl-2,3-dihydro-1h-indole-1-carboxylate, 1,1-dimethylethyl 2,3-dihydro-2 hydroxymethyl-1h-indole-1-carboxylate, 1,1-dimethylethyl 2,3-dihydro-2-hydroxymethyl-1h-indole-1-carboxylate
PubChem CID 2794669
IUPAC Name tert-butyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)CO

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 249.31
Percent Purity 97%
Chemical Name or Material tert-Butyl 2-(hydroxymethyl)-1-indolinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.