missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-bromobenzylcarbamate, ≥97%, Thermo Scientific™

Product Code. 10573583
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10573583 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10573583 Supplier Thermo Scientific Maybridge Supplier No. MO07557DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 162356-90-3
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H16BrNO2
MDL Number MFCD05664385
Quantity 1 g
Synonym tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804
InChI Key DFNZFCPEUDSNEO-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NCC1=CC=CC=C1Br
IUPAC Name tert-butyl N-[(2-bromophenyl)methyl]carbamate
Molecular Weight (g/mol) 286.169
PubChem CID 7018300
Formula Weight 286.17
Percent Purity 97%
Chemical Name or Material Tert-butyl 2-bromobenzylcarbamate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.