missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-aminoisobutyrate hydrochloride, 98%

Product Code. 15420719
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420719 5 g 5g
15478536 250 mg 250mg
15410719 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420719 Supplier Thermo Scientific Alfa Aesar Supplier No. H63817.06
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 84758-81-6 | C8H17NO2 | 159.23 g/mol

tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.

Solubility
Soluble in water or 1% acetic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84758-81-6
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00237698
InChI Key LSVYCJILORYVCD-UHFFFAOYSA-N
Synonym alpha-aminoisobutyric acid t-butyl ester, h-aib-otbu.hcl, tert-butyl 2-methylalaninate, 2-aminoisobutyric acid t-butyl ester, alanine, 2-methyl-, 1,1-dimethylethyl ester, 2-amino-2-methylpropionic acid tert-butyl ester, h-aib-otbu inverted exclamation mark currencyhcl, t-butyl 2-methylalaninate, 2-methylalanine t-butyl ester, h-alpha-me-ala-otbu . hcl
PubChem CID 7019940
IUPAC Name tert-butyl 2-amino-2-methylpropanoate
SMILES CC(C)(C)OC(=O)C(C)(C)N

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in water or 1% acetic acid.
Formula Weight 195.69
Percent Purity 98%
Physical Form Powder
Chemical Name or Material tert-Butyl 2-aminoisobutyrate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.