missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-amino-2-thioxoethylcarbamate, 90%, Thermo Scientific™

Product Code. 10051674
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051674 10 g Amber Glass Bottle 10g
10443245 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10051674 Supplier Thermo Scientific Maybridge Supplier No. MO07925EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89226-13-1
Molecular Formula C7H14N2O2S
Molecular Weight (g/mol) 190.261
MDL Number MFCD09025922
InChI Key AGBIUUFZUPNDTM-UHFFFAOYSA-N
Synonym tert-butyl 2-amino-2-thioxoethyl carbamate, tert-butyl 2-amino-2-thioxoethylcarbamate, tert-butyl n-carbamothioylmethyl carbamate, tert-butyl n-2-amino-2-sulfanylideneethyl carbamate, carbamic acid, 2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, acmc-20ljf8, n-tert-butoxycarbonylglycinethioamide, tert-butoxycarbonylamino acetothioamide, n-t-butoxycarbonylamino acetothioamide
PubChem CID 5324304
IUPAC Name tert-butyl N-(2-amino-2-sulfanylideneethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC(=S)N

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 190.27
Percent Purity 90%
Chemical Name or Material Tert-butyl 2-amino-2-thioxoethylcarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.