missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (2,6-Dioxopiperidin-3-yl)carbamate, TRC

Product Code. 30331464
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331464 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331464 Supplier Toronto Research Chemicals Supplier No. TRCB69239510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl (2,6-Dioxopiperidin-3-yl)carbamate
CAS 31140-42-8
Molecular Formula C10H16N2O4
Quantity 10 g
Synonym tert-Butyl Ester 2,6-Dioxo-3-piperidinecarbamic Acid, 1,1-Dimethylethyl Ester (2,6-Dioxo-3-piperidinyl)-carbamic Acid, Pomalidomide Impurity
InChI Formula InChI=1S/C10H16N2O4/c1-10(2,3)16-9(15)11-6-4-5-7(13)12-8(6)14/h6H,4-5H2,1-3H3,(H,11,15)(H,12,13,14)
SMILES CC(C)(C)OC(=O)NC1CCC(=O)NC1=O
IUPAC Name tert-butyl N-(2,6-dioxopiperidin-3-yl)carbamate
Molecular Weight (g/mol) 228.25
Formula Weight 228.111
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.