missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2,2,2-trichloroacetimidate, 95%

Product Code. 10415053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10415053 25 g Glass bottle 25g
10767403 5 g Glass bottle 5g
11905601 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10415053 Supplier Thermo Scientific Acros Supplier No. 369330250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98946-18-0 | C6H10Cl3NO | 218.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98946-18-0
Molecular Formula C6H10Cl3NO
Molecular Weight (g/mol) 218.50
MDL Number MFCD00077410
InChI Key CQXDYHPBXDZWBA-UHFFFAOYSA-N
Synonym tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine
PubChem CID 2734700
IUPAC Name tert-butyl 2,2,2-trichloroethanimidate
SMILES CC(C)(C)OC(=N)C(Cl)(Cl)Cl

Specifications

Melting Point 20.0°C
Color Colorless to Yellow
Density 1.2220g/mL
Boiling Point 64.0°C to 66.0°C (15.0 mmHg)
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CCl3C(=NH)OC(CH3)3
Refractive Index 1.4550 to 1.4570
Quantity 25 g
Specific Gravity 1.222
Formula Weight 218.51
Percent Purity 95%
Physical Form Liquid or Low Melting Crystalline Mass
Chemical Name or Material tert-Butyl 2, 2, 2-trichloroacetimidate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl 2,2,2-trichloroacetimidate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.